HUVEC – Human Umbilical Vein Endothelial Cells, Pooled, in EGM®-2
Supplier: LONZA PHARMA - BIOSCIENCE CA
C2519A
CA10064-186EA 612.25 CAD
CA10064-186
HUVEC – Human Umbilical Vein Endothelial Cells, Pooled, in EGM®-2
Cells
Lonza's cryopreserved HUVECs are shipped as frozen primaries. They are guaranteed to express CD31/105 and survive in culture through 15* population doublings (*except HUVEC-XL® guaranteed through 5 PD).
  • Guaranteed through 15 population doublings
  • Test negative for mycoplasma, bacteria, yeast, and fungi
  • HIV-1, Hepatitis B and Hepatitis C are not detected for all donors and/or cell lots
  • A Certificate of Analysis is provided for each cell lot purchased
Our pooled vs. single donor options allow researchers to study or exclude donor variability from their experiments. Depending upon the study, pooled version is typically utilized to reduce donor variability while single donor option allows researchers to assess donor-specific differences.
HUVEC-XL size consists of 10 million cells/amp achieved by pooling donors. This option is designed to support studies that require large volume of cells while still offering cost-benefits to researchers.
Suitable HUVECs for research:
C2517A (single donor), C2519A (pooled donor), 00191027 (XL size) - Isolated in EGM®-2 media, in presence of growth factors such as VEGF, rhFGF, rhEGF, and others. Does not contain BBE.
C2517AS (single donor), C2519AS (pooled donor) - Pre-screened HUVECs in EGM®-2 media. Screened for angiogenesis markers and test positive for expression of eNOS, Axl, Tie-2 and VEGFr2.
CC-2517 (single donor), CC-2519 (pooled donor) - Both isolated in EGM® media, in absence of defined growth factors. Contains BBE. No exogenous VEGF.
CC-2935 (single donor) - Isolated in EGM® - Plus media: Improved version of part number CC-2517. Offers faster proliferation rates than HUVEC isolated in EGM® Media. No exogenous VEGF.
All cells test negative for mycoplasma, bacteria, yeast, and fungi. HIV-1, hepatitis B and hepatitis C are not detected for all donors and/or cell lots
Applications: Wound healing, angiogenesis, tissue engineering, inflammation, oncology, pharmacology, vascular modeling, transfection.
Delivery information: Content: 1×Cryopreserved ampule of HUVEC containing ≥500000 cells.