KBM® Gold Keratinocyte Growth Basal Medium, Calcium and Phenol Red Free
Supplier: LONZA PHARMA - BIOSCIENCE CA
00195130
CA10063-830EA 207.38 CAD
CA10063-830
KBM® Gold Keratinocyte Growth Basal Medium, Calcium and Phenol Red Free
Cell Culture Media
KBM® Gold is a calcium-free keratinocyte basal medium sold separately. This can be used with 1 kit of keratinocyte cell growth medium SingleQuots® Supplements and Growth Factors.
KGM® Gold BulleKit® is a new robust and versatile serum-free medium formulation for high and low density NHEK-Normal Human Epidermal Keratinocyte cultures.
Delivery information: Content: 1×bottle basal medium 500 ml.