Amersham™ Protran® Premium Western Blotting Membranes, Nitrocellulose, Cytiva
Supplier: Cytiva
10600008 10600003 10600004 10600013 10600014 10600034 10600047 10600048 10600065 10600079 10600096 10600009 10600117 10600118 10600080
CA10061-088PK 975.38 CAD
CA10061-088 CA10061-120 CA10061-122 CA10061-128 CA10061-130 CA10061-218 CA10061-234 CA10061-236 CA10061-250 CA10061-272 CA10061-274 CA10061-316 CA10061-334 CA10061-336 CA10061-394 MPCA29489442
Amersham™ Protran® Premium Western Blotting Membranes, Nitrocellulose, Cytiva
Transfer Membranes
Amersham™ Protran® Premium nitrocellulose blotting membranes provide excellent sensitivity, resolution and low background in fluorescence Western blotting detection applications.
  • Available in popular pre-cut sizes
  • High binding capacity
  • Protran® Premium 0.2 NC offers low auto-fluorescence across a wide range of excitation/emission wavelengths, making the membrane suitable for fluorescence-based immunodetection and multiplexing
  • Protran® Premium 0.45 NC is an unsupported, 100% pure nitrocellulose membrane suitable for proteins of a wide range of molecular
More Product Information