Amersham™ Protran® Western Blotting Membranes, Nitrocellulose, Cytiva
Supplier: Cytiva
10600006 10600007 10600000 10600001 10600002 10600005 10600012 10600032 10600033 10600041 10600042 10600043 10600044 10600045 10600046 10600062 10600073 10600010 10600011 10600114 10600115 10600116 10600093 10600094 10600124 10600125
CA10061-084PK 768.12 CAD
CA10061-084 CA10061-086 CA10061-090 CA10061-092 CA10061-094 CA10061-124 CA10061-126 CA10061-214 CA10061-216 CA10061-222 CA10061-224 CA10061-226 CA10061-228 CA10061-230 CA10061-232 CA10061-244 CA10061-260 CA10061-318 CA10061-320 CA10061-328 CA10061-330 CA10061-332 CA10061-406 CA10061-408 CA10143-074 CA10143-076 MPCA29489439
Amersham™ Protran® Western Blotting Membranes, Nitrocellulose, Cytiva
Transfer Membranes
Amersham™ Protran® membranes are manufactured from 100% pure nitrocellulose to ensure the highest binding capacity possible. They are compatible with a variety of detection methods, including isotopic, chemiluminescent, colorimetric and fluorescent. No methanol pre-wetting step is required, making this the membrane of choice for proteins which prefer aqueous environments. Prior to transfer the membrane is simply wetted with water, and then placed in the transfer buffer. No other pretreatment steps are necessary.
  • Proprietary nitrocellulose formula gives a long shelf life of bound
More Product Information