Sterile Serological Pipettes
Supplier: Eppendorf
0030127692 0030127706 0030127714 0030127722 0030127730 0030127749 0030127730
CA10054-532EA 320.02 CAD
CA10054-532 CA10054-534 CA10054-536 CA10054-538 CA10054-540 CA10054-542 10048-947
Sterile Serological Pipettes
Pipettes Serological Pipettes
Eppendorf Serological Pipettes ensure top-tier sterility and safety for sensitive lab applications. They are sterile, free from pyrogens, DNase, RNase, human and bacterial DNA, and non-cytotoxic. Ideal for cell culture and molecular biology tasks.
  • Clear and precise graduations for easy volume determination
  • Color-coded for easy identification of pipet sizes
  • Subunits with dispenser option to keep stored products safe
  • Individually wrapped for reliable sterility
  • Ultra-pure virgin polystyrene meets the requirement of USP VI
  • Harmonized and well-fitting operation
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable