Anti-CPDA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemical
200-4352S
CA10037-070EA 229.01 CAD
CA10037-070
Anti-CPDA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Carboxypeptidase A is useful in ELISA, western blot, and immunochemistry. Optimal titers are to be reached by researchers.

Recommended Dilutions: ELISA: 1:3000 - 1:10000; Western Blot: 1:300 - 1:2000;
Do not add Sodium Azide!

Type: Primary
Antigen: CPDA
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR