Certified Reference Materials, Oxide, Rocklabs
Supplier: ROCKLABS (W/T) CA
OXP-AU-2.5KG OXJ-AU-2.5KG OXK-AU-2.5KG OXD-AU-2.5KG OXC-AU-2.5KG OXG-AU-2.5KG OXQ-AU-2.5KG OXH-AU-2.5KG OXI-AU-2.5KG OXA-AU-2.5KG OXE-AU-2.5KG OXF-AU-2.5KG OXL-AU-2.5KG OXN-AU-2.5KG OXA-AU-100X100G OXC-AU-100X100G OXD-AU-100X100G OXE-AU-100X100G OXF-AU-100X100G OXG-AU-100X100G OXH-AU-100X100G OXI-AU-100X100G OXK-AU-100X100G OXL-AU-100X100G OXP-AU-100X100G OXA-AU-100X30G OXC-AU-100X30G OXD-AU-100X30G OXE-AU-100X30G OXH-AU-100X30G OXI-AU-100X30G OXJ-AU-100X30G OXK-AU-100X30G OXP-AU-100X50G OXQ-AU-100X50G OXF-AU-100X30G OXG-AU-100X30G OXL-AU-100X30G OXN-AU-100X30G OXP-AU-100X30G OXQ-AU-100X30G OXA-AU-100X50G OXC-AU-100X50G OXD-AU-100X50G OXE-AU-100X50G OXF-AU-100X50G OXG-AU-100X50G OXH-AU-100X50G OXI-AU-100X50G OXJ-AU-100X50G OXK-AU-100X50G OXL-AU-100X50G OXN-AU-100X50G OXB-AU-2.5KG
CA10029-286EA 380.88 CAD
CA10029-286 CA10032-320 CA10032-322 CA10032-610 CA10032-614 CA10032-616 CA10032-618 CA10032-620 CA10032-622 CA10067-780 CA10067-782 CA10067-784 CA10067-786 CA10067-788 CA10067-792 CA10067-794 CA10067-796 CA10067-798 CA10067-802 CA10067-806 CA10067-810 CA10067-814 CA10067-820 CA10067-824 CA10067-830 CA10067-836 CA10067-838 CA10067-840 CA10067-842 CA10067-846 CA10067-850 CA10067-854 CA10067-858 CA10067-864 CA10067-868 CA10067-872 CA10067-876 CA10067-880 CA10067-884 CA10067-888 CA10067-892 CA10067-896 CA10067-898 CA10067-900 CA10067-902 CA10067-906 CA10067-910 CA10067-914 CA10067-918 CA10067-922 CA10067-926 CA10067-930 CA10067-934 CA10171-190
Certified Reference Materials, Oxide, Rocklabs
Standards Environmental Analysis Standards
Rocklabs Gold Standards are reference materials that are certified for either Pt, Pd, Au or Ag
Recommended Gold (ppm) concentrations. Packaged in jars or satchets.
Order Now
SPECIFICATIONS
Environmentally Preferable