Terrific Broth with 1.6% Glycerol Bottled Media, Teknova

Supplier: TEKNOVA CA

T7100
CA100219-892EA 368.82 CAD
CA100219-892
Terrific Broth with 1.6% Glycerol Bottled Media, Teknova
Media for Microbiology Prepared Media for Microbiology

Used to obtain high yields of plasmid DNA from E.coli.


  • Higher plasmid yields may be obtained than with standard terrific broth
  • Sterile


Add 100mL of 10X phosphate buffer to 900mL of terrific broth


Contains 1.2% Tryptone, 2.4% yeast extract,1.6% glycerol. Ambient shipping conditions. Store at room temperature.


The 100mL of 10X phosphate buffer supplement is supplied separately (P7100).

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR