LB Agar Plates with 100 µg/ml Ampicillin, X-gal and IPTG, Teknova
Supplier: TEKNOVA CA
L1902
CA100219-318PK 1284.43 CAD
CA100219-318
LB Agar Plates with 100 µg/ml Ampicillin, X-gal and IPTG, Teknova
Media for Microbiology Prepared Media for Microbiology
For use with ampicillin resistant strains and strains harboring plasmids such as pBluescript, pGEM, pUC series plasmids. X-gal allows blue/white selection of colonies.
  • Sleeve of 20 plates
  • Sterile
Contains 1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% agar 100 µg/ml ampicillin, 0.1 mM IPTG, 60 µg/ml X-gal.
Ambient shipping conditions. Refrigerate to store.