Teknova Growth Media and Solutions for Manipulating Bacteriophage Lambda, Teknova
Supplier: TEKNOVA CA
M1260 M1200 Y4220 M1201 Y4221 Y4010 Y4210 M1261
CA100216-346PK 782.94 CAD
CA100216-346 CA100216-636 CA100216-658 CA100216-916 CA100217-442 CA100219-022 CA100219-192 CA100219-360 CA100219-564 CA100219-782
Teknova Growth Media and Solutions for Manipulating Bacteriophage Lambda, Teknova
Media for Microbiology Prepared Media for Microbiology
With proper storage, most antibiotic plates are useable for up to two months.
  • Shelf-life dated
  • Useable for up to two months
Plates are shelf-life dated and guaranteed for 4 weeks from the date of shipment. All plates must be stored at 4°C and in the absence of continuous exposure to light. With proper storage, most antibiotic plates are useable for up to 2 months. All plates sold in sleeves of 20.