CANNON® CL Low Temperature Viscosity Standards, Cannon Instruments
Supplier: CANNON INSTRUMENT COMPANY CA
9727-N02.016 9727-N02.032 9727-N02.128 9727-N02.500 9727-N04.016 9727-N04.032 9727-N04.128 9727-N04.500 9727-N06.016 9727-N06.032 9727-N06.128 9727-N06.500 9727-N07.016 9727-N07.032 9727-N07.128 9727-N07.500 9727-N08.016 9727-N08.032 9727-N08.128 9727-N08.500 9727-N09.128 9727-N09.500 9727-N10.016 9727-N10.032 9727-N10.128 9727-N10.500 9727-N11.016 9727-N11.032 9727-N11.128 9727-N11.500 9727-N12.016 9727-N12.032 9727-N12.128 9727-N12.500 9727-N13.016 9727-N13.032 9727-N13.128 9727-N13.500 9727-N16.032 9727-N16.128 9727-N16.500 9727-N18.016 9727-N18.032 9727-N18.128 9727-N18.500 9727-N20.016 9727-N20.032 9727-N20.128 9727-N20.500 9727-N22.016 9727-N22.032 9727-N22.128 9727-N22.500 9727-N24.016 9727-N24.032 9727-N24.128 9727-N24.500 9727-N26.016 9727-N26.032 9727-N26.128 9727-N26.500 9727-N28.016 9727-N28.032 9727-N28.128 9727-N28.500 9727-N30.016 9727-N31.016 9727-N31.032 9727-N31.128 9727-N31.500 9727-N32.016 9727-N32.032 9727-N32.128 9727-N32.500 9727-N33.016 9727-N34.016 9727-N34.032 9727-N34.128 9727-N34.500 9727-N35.016 9727-N35.032 9727-N35.128 9727-N35.500 9727-N36.016 9727-N36.032 9727-N36.128 9727-N36.500 9727-N37.016 9727-N37.032 9727-N37.128 9727-N37.500 9727-N16.016 9727-N33.032 9727-N33.128 9727-N33.500 9727-N09.016 9727-N09.032 9727-N30.032 9727-N30.128 9727-N30.500 9727-N14.016 9727-N14.032 9727-N14.128 9727-N14.500
CA10021-474EA 216.4 CAD
CA10021-474 CA10021-476 CA10021-478 CA10021-480 CA10021-484 CA10021-486 CA10021-488 CA10021-490 CA10021-492 CA10021-494 CA10021-496 CA10021-498 CA10021-502 CA10021-504 CA10021-506 CA10021-508 CA10021-510 CA10021-512 CA10021-514 CA10021-516 CA10021-518 CA10021-520 CA10021-522 CA10021-524 CA10021-526 CA10021-528 CA10021-532 CA10021-534 CA10021-536 CA10021-538 CA10021-540 CA10021-542 CA10021-544 CA10021-546 CA10021-550 CA10021-552 CA10021-554 CA10021-556 CA10021-558 CA10021-560 CA10021-562 CA10021-564 CA10021-566 CA10021-568 CA10021-570 CA10021-574 CA10021-576 CA10021-578 CA10021-580 CA10021-582 CA10021-584 CA10021-586 CA10021-588 CA10021-592 CA10021-594 CA10021-596 CA10021-598 CA10021-600 CA10021-602 CA10021-604 CA10021-606 CA10021-610 CA10021-612 CA10021-614 CA10021-616 CA10021-618 CA10021-622 CA10021-624 CA10021-626 CA10021-628 CA10021-630 CA10021-632 CA10021-634 CA10021-636 CA10021-640 CA10021-642 CA10021-644 CA10021-646 CA10021-648 CA10021-652 CA10021-654 CA10021-656 CA10021-658 CA10021-660 CA10021-662 CA10021-664 CA10021-666 CA10021-670 CA10021-672 CA10021-674 CA10021-676 CA10021-680 CA10021-684 CA10021-996 CA10021-998 CA10022-000 CA10022-002 CA10022-058 CA10022-060 CA10022-062 CA10022-064 CA10022-066 CA10022-114 CA10022-116 CA10022-118 CA10022-120
CANNON® CL Low Temperature Viscosity Standards, Cannon Instruments
Standards Viscometry Standards
CANNON® CL standards provide certified dynamic viscosity data at temperatures from –5 °C to –35 °C. These standards are used to calibrate the cold-cranking simulator for oil testing.
Certifications: Certified to ASTM D 5293 and SAE Specification J300.