General Purpose Viscosity Standards, Cannon Instruments
Supplier: CANNON INSTRUMENT COMPANY CA
9727-C40.016 9727-C35.016 9727-C10.016 9727-C15.016 9727-C20.016 9727-C22.016 9727-C25.016 9727-C25.032 9727-C25.128 9727-C25.500 9727-C27.016 9727-C27.032 9727-C27.128 9727-C27.500 9727-C30.016 9727-C30.032 9727-C30.128 9727-C30.500 9727-C31.016 9727-C31.032 9727-C31.128 9727-C34.500 9727-C35.032 9727-C35.128 9727-C35.500 9727-C36.016 9727-C36.032 9727-C36.128 9727-C36.500 9727-C37.016 9727-C37.032 9727-C37.128 9727-C37.500 9727-C38.016 9727-C38.032 9727-C38.128 9727-C38.500 9727-C40.016 9727-C40.032 9727-C41.016 9727-C42.500 9727-C43.016 9727-C43.032 9727-C43.128 9727-C43.500 9727-C45.016 9727-C45.032 9727-C45.128 9727-C45.500 9727-C46.016 9727-C46.032 9727-C46.128 9727-C46.500 9727-C47.016 9727-C47.032 9727-C47.128 9727-C47.500 9727-C48.016 9727-C48.032 9727-C48.128 9727-C48.500 9727-C50.016 9727-C50.032 9727-C50.128 9727-C50.500 9727-C51.016 9727-C51.032 9727-C51.128 9727-C51.500 9727-C52.016 9727-C52.032 9727-C52.128 9727-C42.016 9727-C42.032 9727-C42.128
CA47735-110EA 812.28 CAD
CA47735-110 CA10215-522 CA10021-118 CA10021-120 CA10021-122 CA10021-126 CA10021-134 CA10021-136 CA10021-138 CA10021-140 CA10021-144 CA10021-146 CA10021-148 CA10021-150 CA10021-152 CA10021-154 CA10021-156 CA10021-158 CA10021-162 CA10021-164 CA10021-166 CA10021-168 CA10021-172 CA10021-174 CA10021-176 CA10021-180 CA10021-182 CA10021-184 CA10021-186 CA10021-188 CA10021-190 CA10021-192 CA10021-194 CA10021-198 CA10021-200 CA10021-202 CA10021-204 CA10021-206 CA10021-208 CA10021-210 CA10021-212 CA10021-216 CA10021-218 CA10021-220 CA10021-222 CA10021-224 CA10021-226 CA10021-228 CA10021-230 CA10021-234 CA10021-236 CA10021-238 CA10021-240 CA10021-242 CA10021-244 CA10021-246 CA10021-248 CA10021-252 CA10021-254 CA10021-256 CA10021-258 CA10021-260 CA10021-262 CA10021-264 CA10021-266 CA10021-270 CA10021-272 CA10021-274 CA10021-276 CA10021-278 CA10021-280 CA10021-282 CA10022-026 CA10022-028 CA10022-030
General Purpose Viscosity Standards, Cannon Instruments
Standards Viscometry Standards
These certified standards are useful for calibration and verification procedures.
  • It is suitable for all types of viscometers
  • It also helps ensures the reliability of measurements
Certifications: Meets the precision specifications of ASTM D 445/446 and ISO 3104/3105, and are traceable to the NIST.