Orion™ Sure-Flow® Combination pH Electrode 9165BNWP, Thermo Scientific
Supplier: Thermo Fisher Scientific
9165BNWP
CA10010-796EA 1678.21 CAD
CA10010-796
Orion™ Sure-Flow® Combination pH Electrode 9165BNWP, Thermo Scientific
Electrodes pH Electrodes
Combination pH electrode with epoxy body for fast response in difficult samples and soils.
  • Refillable Ag/AgCl combination pH electrode
  • Waterproof BNC connector
Compatible with Orion Star™ Series meters and competitive meter models with BNC connection.