DuPont™ Tychem® 2000 Coveralls with Standard Hood and Elastic Wrists and Ankles, Bound Seams
Supplier: DuPont
QC127BYLMD001200 QC127BYLLG001200 QC127BYLXL001200 QC127BYL2X001200 QC127BYL3X001200 QC127BYL4X001200 QC127BYL5X001200 QC127BYL6X001200
CA10010-252CS 1318.82 CAD
CA10010-252 CA10010-254 CA10010-256 CA10010-258 CA10010-260 CA10010-262 10016-404 10016-406
DuPont™ Tychem® 2000 Coveralls with Standard Hood and Elastic Wrists and Ankles, Bound Seams
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Standard fit hood
  • Elastic wrists and ankles
  • Storm flap with adhesive closure
  • Bound seams
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Tightly sewn seam isMore Product Information