DuPont™ Tychem® 2000 Coveralls with Standard Hood and Attached Socks, Bound Seams
Supplier: DuPont
QC122BYLMD001200 QC122BYLXL001200 QC122BYL2X001200 QC122BYL3X001200 QC122BYL4X001200 QC122BYL2X0012BN QC122BYL3X0012BN QC122BYL4X0012BN QC122BYL5X0012BN QC122BYL6X001200 QC122BYL6X0012BN QC122BYLLG0012BN QC122BYLMD0012BN QC122BYLXL0012BN
CA10010-238CS 1412.26 CAD
CA10010-238 CA10010-240 CA10010-242 CA10010-244 CA10010-246 CA10010-248 10016-370 10016-372 10016-374 10016-376 10016-378 10016-380 10016-382 10016-384 10016-386
DuPont™ Tychem® 2000 Coveralls with Standard Hood and Attached Socks, Bound Seams
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Standard fit hood
  • Elastic wrists
  • Attached socks
  • Storm flap with adhesive closure
  • Bound seams
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Tightly sewnMore Product Information