DuPont™ Tyvek® 400 Frocks with Laydown Collar
Supplier: DuPont
TY210SWHMD003000 TY210SWHXL003000 TY210SWHSM003000 TY210SWHLG003000 TY210SWH2X003000 TY210SWH3X003000 TY210SWH4X003000 TY210SWH5X003000 TY210SWH6X003000 TY210SWH7X003000
CA10010-070CS 1305.83 CAD
CA10010-070 CA10010-074 CA10010-068 CA10010-072 CA10010-076 CA10010-078 CA10010-080 10016-918 10016-920 10016-922
DuPont™ Tyvek® 400 Frocks with Laydown Collar
Disposable Garments
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Laydown collar
  • Open wrists
  • Extends below hips
  • Front snap closure
  • Serged seams
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durabilityMore Product Information