HyFlex® 11-541 Ultralight Weight, 18-Gauge Gloves, Palm Dipped, Ansell
Supplier: Ansell Canada
11541060 11541070 11541080 11541090 11541100 11541110
CA10002-004CS 5477.12 CAD
CA10002-004 CA10002-074 CA10002-076 CA10002-078 CA10002-080 CA10002-082
HyFlex® 11-541 Ultralight Weight, 18-Gauge Gloves, Palm Dipped, Ansell
Gloves
Offering exceptional cut protection in a form-fitting ultralight weight glove with enhanced grip performance, the HyFlex® 11-541 epitomizes the convergence of protection and performance.
  • Enhanced abrasion resistance
  • Excellent grip
  • Superior tactile sensitivity
  • Unparalleled dexterity
It features an 18G ultra-lightweight liner with INTERCEPT TECHNOLOGY® yarns that provides EN Cut Level 4 (ANSI Cut Level 4) protection.
This glove's ultralight weight enables unparalleled dexterity and refined mobility for bare hand-like tactility. ZONZ™ comfort fitMore Product Information