MycoSeal™ Shrink Seals, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
SS9225
CA10001-500EA 204.75 CAD
CA10001-500
MycoSeal™ Shrink Seals, Hardy Diagnostics
Seals Security Seals
Cellulose shrink seals are recommended for sealing petri plates.
  • Cellulose shrink seal
  • Designed to keep petri plate lids tightly sealed
  • Prevents spores from becoming airborne
Cellulose shrink seals are recommended for sealing 15x100mm petri plates. It is a cellulose shrink seal designed to keep petri plate lids tightly sealed. Sealed plates retain their moisture and stay fresh longer. Ideal for mycology and TB cultures, MycoSeal™ prevents dehydration and cracking of the media and yet still allows both oxygen and carbon dioxide gas exchange. The tightMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable