HiTrap Columns, Capto Phenyl ImpRes
Supplier: Cytiva
17-5484-11 17548412
CA10000-218EA 973.83 CAD
CA10000-218 CA77509-282
HiTrap Columns, Capto Phenyl ImpRes
Chromatography Columns
HiTrap Capto Phenyl ImpRes is prepacked with Capto Phenyl ImpRes media for screening and small-scale protein purifications using hydrophobic interaction chromatography.
  • High-throughput medium for intermediate and polishing protein purification, excellent choice for process-scale purification
  • Columns for screening of selectivity, binding and elution conditions, as well as small scale purifications
  • Excellent chemical stability allows cleaning with all standard CIP solutions, including 1 M sodium hydroxide
The combined properties of high binding capacity,More Product Information