Anti-CIITA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-401-249
CA100-401-249EA 834 CAD
CA100-401-249
Anti-CIITA Rabbit Polyclonal Antibody
Antibodies
For immunoblotting a 1:500 dilution is recommended. Researchers should determine optimal titers for other applications.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;

Type: Primary
Antigen: CIITA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR