Samarium Standard for ICP, Certipur®, MilliporeSigma
Supplier: MilliporeSigma
1.70348.0100
CA1.70348.0100CS 15137 CAD
CA1.70348.0100
Samarium Standard for ICP, Certipur®, MilliporeSigma
Standards Cationic Standards Samarium Standards
Sm₂O₃ in HNO₃
Traceable to SRM from NIST.
Certifications: The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Ordering information: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.
Order Now
SPECIFICATIONS
Environmentally Preferable
Hazard class description Corrosive Material