LiChroCART® Stainless Steel LiChrospher® Cartridges, MilliporeSigma
Supplier: MilliporeSigma
1.50148.0001 1.50822.0001 1.50829.0001 1.50834.0001 1.50943.0001 1.50154.0001 1.50158.0001 1.50987.0001 1.50958.0001 1.50961.0001 1.50963.0001 1.50983.0001 1.50742.0001 1.50826.0001 1.50833.0001 1.50837.0001 1.50843.0001 1.50847.0001 1.50931.0001 1.50937.0001 1.50984.0001 1.50742.0001 1.50822.0001 1.50824.0001 1.50826.0001 1.50828.0001 1.50829.0001 1.50833.0001 1.50834.0001 1.50837.0001 1.50842.0001 1.50843.0001 1.50847.0001 1.50154.0001 1.50158.0001 1.50159.0001 1.50987.0001 1.50963.0001 1.50148.0001 1.50931.0001 1.50937.0001 1.50943.0001 1.50958.0001 1.50961.0001 1.50983.0001 1.50984.0001 1.50155.0001 1.50600.0001 1.50601.0001 1.50602.0001 1.50603.0001 1.50605.0001 1.50635.0001 1.50636.0001 1.50638.0001 1.50639.0001 1.50641.0001 1.50642.0001 1.50840.0001 1.50858.0001
48219-312PK 761.56 CAD
48219-312 48219-368 48219-382 48219-390 48219-456 48219-508 48219-514 48219-562 48219-582 48219-588 48219-592 48219-616 48219-362 48219-376 48219-388 48219-394 48219-404 48219-410 48219-442 48219-450 48219-618 CA48219-362 CA48219-368 CA48219-372 CA48219-376 CA48219-380 CA48219-382 CA48219-388 CA48219-390 CA48219-394 CA48219-402 CA48219-404 CA48219-410 CA48219-508 CA48219-514 CA48219-516 CA48219-562 CA48219-592 CA48219-312 CA48219-442 CA48219-450 CA48219-456 CA48219-582 CA48219-588 CA48219-616 CA48219-618 CA1.50155.0001 CA1.50600.0001 CA1.50601.0001 CA1.50602.0001 CA1.50603.0001 CA1.50605.0001 CA1.50635.0001 CA1.50636.0001 CA1.50638.0001 CA1.50639.0001 CA1.50641.0001 CA1.50642.0001 CA1.50840.0001 CA1.50858.0001 CA1.50928.0001
LiChroCART® Stainless Steel LiChrospher® Cartridges, MilliporeSigma
Chromatography Columns
Cartridges are filled with reliable, versatile, traditionally produced spherical silica carrier materials.
RP-18, RP-18E (Endcapped), RP-8, and RP-8E (Endcapped) cartridges are ideal for reproducible reversed phase separation in chromatography applications including acidic, neutral, and weakly basic compounds. RP-Select B cartridges can be used to provide excellent separations of basic compounds and neutral or acidic substances. Si (silica gel) cartridges are ideal for normal phase HPLC applications. CN (cyano) cartridges are ideal for chromatography applications with complex samples withMore Product Information