SeQuant® ZIC®-HILIC HPLC Columns, MilliporeSigma
Supplier: MilliporeSigma
1.50447.0001 1.50451.0001 1.50489.0001 1.50490.0001 1.50447.0001 1.50451.0001 1.50489.0001 1.50490.0001 1.50671.0001 1.50671.0001 1.50435.0001 1.50440.0001 1.50441.0001 1.50442.0001 1.50443.0001 1.50444.0001 1.50445.0001 1.50446.0001 1.50448.0001 1.50449.0001 1.50450.0001 1.50452.0001 1.50453.0001 1.50454.0001 1.50455.0001 1.50457.0001 1.50458.0001 1.50465.0001 1.50478.0001 1.50479.0001 1.50480.0001 1.50481.0001 1.50482.0001 1.50487.0001 1.50492.0001 1.50493.0001 1.50494.0001 1.50496.0001 1.50497.0001
EM1.50447.0001EA 2089.08 CAD
EM1.50447.0001 EM1.50451.0001 EM1.50489.0001 EM1.50490.0001 CA1.50447.0001 CA1.50451.0001 CA1.50489.0001 CA1.50490.0001 52428-712 CA1.50671.0001 CA1.50434.0001 CA1.50435.0001 CA1.50440.0001 CA1.50441.0001 CA1.50442.0001 CA1.50443.0001 CA1.50444.0001 CA1.50445.0001 CA1.50446.0001 CA1.50448.0001 CA1.50449.0001 CA1.50450.0001 CA1.50452.0001 CA1.50453.0001 CA1.50454.0001 CA1.50455.0001 CA1.50457.0001 CA1.50458.0001 CA1.50465.0001 CA1.50478.0001 CA1.50479.0001 CA1.50480.0001 CA1.50481.0001 CA1.50482.0001 CA1.50487.0001 CA1.50492.0001 CA1.50493.0001 CA1.50494.0001 CA1.50496.0001 CA1.50497.0001
SeQuant® ZIC®-HILIC HPLC Columns, MilliporeSigma
Chromatography Columns
The selectivity offered by ZIC®-HILIC (Hydrophilic Interaction Liquid Chromatography) is suitable for a wide variety of molecules containing hydrophilic or ionisable functional groups.
  • Straightforward separation of polar and hydrophilic compounds
  • Orthogonal selectivity compared to reversed-phase
  • High-density bonded sulfobetaine zwitterionic stationary phase
  • Low cost per analysis due to robust stationary phase
This includes compounds such as carbohydrates, metabolites, acids and bases, organic and inorganic ions, metal complexes, amino acids, peptides,More Product Information