Purospher® STAR HPLC Columns and Cartridges, MilliporeSigma
Supplier: MilliporeSigma
1.50244.0001 1.50245.0001 1.50248.0001 1.50250.0001 1.50251.0001 1.50252.0001 1.50267.0001 1.50358.0001 1.50225.0001 1.50231.0001 1.50240.0001 1.50241.0001 1.50242.0001 1.50249.0001 1.50268.0001 1.50356.0001 1.50357.0001 1.50272.0001 1.50031.0001 1.50032.0001 1.50033.0001 1.50035.0001 1.51453.0001 1.51454.0001 1.50036.0001 1.50037.0001 1.51455.0001 1.50274.0001 1.50038.0001 1.50255.0001 1.50253.0001 1.51460.0001 1.50253.0001 1.50244.0001 1.50245.0001 1.50247.0001 1.50248.0001 1.50250.0001 1.50251.0001 1.50252.0001 1.50267.0001 1.50358.0001 1.50359.0001 1.50225.0001 1.50231.0001 1.50239.0001 1.50240.0001 1.50241.0001 1.50242.0001 1.50249.0001 1.50268.0001 1.50356.0001 1.50357.0001 1.50270.0001 1.50271.0001 1.50272.0001 1.50031.0001 1.50032.0001 1.50033.0001 1.50035.0001 1.51453.0001 1.51454.0001 1.50036.0001 1.50037.0001 1.51455.0001 1.50274.0001 1.50038.0001 1.50255.0001 1.51460.0001 1.50623.0001 1.50624.0001 1.50625.0001 1.50626.0001 1.50627.0001 1.50623.0001 1.50624.0001 1.50625.0001 1.50626.0001 1.50627.0001 1.50254.0001 1.50256.0001 1.51918.0001 1.50750.0001
48219-470EA 1154.36 CAD
48219-470 48219-472 48219-474 48219-476 48219-478 48219-480 48219-482 48219-484 48219-486 48219-488 48219-754 48219-760 48219-768 48219-770 48219-772 48219-782 48219-784 48219-787 48219-789 48219-794 48219-796 48219-798 48219-800 48219-803 48219-805 48219-806 48219-808 48219-810 48219-812 48219-817 48219-821 48219-825 48219-828 48219-870 CA1.50253.0001 CA48219-470 CA48219-472 CA48219-474 CA48219-476 CA48219-478 CA48219-480 CA48219-482 CA48219-484 CA48219-486 CA48219-488 CA48219-754 CA48219-760 CA48219-766 CA48219-768 CA48219-770 CA48219-772 CA48219-782 CA48219-784 CA48219-787 CA48219-789 CA48219-790 CA48219-793 CA48219-794 CA48219-796 CA48219-798 CA48219-800 CA48219-803 CA48219-805 CA48219-806 CA48219-808 CA48219-810 CA48219-812 CA48219-817 CA48219-821 CA48219-825 CA48219-870 EM1.50623.0001 EM1.50624.0001 EM1.50625.0001 EM1.50626.0001 EM1.50627.0001 CA1.50623.0001 CA1.50624.0001 CA1.50625.0001 CA1.50626.0001 CA1.50627.0001 CA1.50254.0001 CA1.50256.0001 CA1.51918.0001 CA1.51919.0001 EM1.50750.0001
Purospher® STAR HPLC Columns and Cartridges, MilliporeSigma
Chromatography Columns
Columns are for use with basic, neutral, metal, chelating, or any other sample format, and are ideal for the separation of difficult hydrophobic samples.
Purospher™ STAR columns are designed to provide optimum results even when loaded with large or small sample volumes and are also available in a narrow-bore format (1 mm). The wide pH range provides the required pH stability for 99% of the common analysis applications. These columns provide the power to separate even the most difficult of compound mixtures. With up to 150000 plates per meter for the 3 µm particle size and 100000 platesMore Product Information