Precoated Glass-Backed TLC Plates, MilliporeSigma
Supplier: MilliporeSigma
1.05434.0001 1.05608.0001 1.05626.0001 1.05628.0001 1.05629.0001 1.05631.0001 1.05632.0001 1.05633.0001 1.05635.0001 1.05641.0001 1.05642.0001 1.05644.0001 1.05713.0001 1.05714.0001 1.05715.0001 1.05716.0001 1.05717.0001 1.05718.0001 1.05719.0001 1.05724.0001 1.05725.0001 1.05728.0001 1.05729.0001 1.05730.0001 1.05731.0001 1.05738.0001 1.05744.0001 1.05745.0001 1.05746.0001 1.05747.0001 1.05786.0001 1.05787.0001 1.05789.0001 1.05794.0001 1.05801.0001 1.05802.0001 1.05804.0001 1.05805.0001 1.05808.0001 1.05914.0001 1.11764.0001 1.11798.0001 1.11844.0001 1.11845.0001 1.11846.0001 1.12668.0001 1.13124.0001 1.13187.0001 1.13724.0001 1.13725.0001 1.13726.0001 1.13727.0001 1.13728.0001 1.13748.0001 1.13792.0001 1.13793.0001 1.13794.0001 1.13894.0001 1.13895.0001 1.15035.0001 1.15036.0001 1.15326.0001 1.15327.0001 1.15341.0001 1.15388.0001 1.15389.0001 1.15423.0001 1.15424.0001 1.15445.0001 1.15552.0001 1.15647.0001 1.15683.0001 1.15684.0001 1.15696.0001 1.16464.0001 1.16485.0001
CA1.05434.0001EA 10203.64 CAD
CA1.05434.0001 CA1.05608.0001 CA1.05626.0001 CA1.05628.0001 CA1.05629.0001 CA1.05631.0001 CA1.05632.0001 CA1.05633.0001 CA1.05635.0001 CA1.05641.0001 CA1.05642.0001 CA1.05644.0001 CA1.05713.0001 CA1.05714.0001 CA1.05715.0001 CA1.05716.0001 CA1.05717.0001 CA1.05718.0001 CA1.05719.0001 CA1.05724.0001 CA1.05725.0001 CA1.05726.0001 CA1.05728.0001 CA1.05729.0001 CA1.05730.0001 CA1.05731.0001 CA1.05738.0001 CA1.05744.0001 CA1.05745.0001 CA1.05746.0001 CA1.05747.0001 CA1.05786.0001 CA1.05787.0001 CA1.05789.0001 CA1.05794.0001 CA1.05801.0001 CA1.05802.0001 CA1.05804.0001 CA1.05805.0001 CA1.05808.0001 CA1.05914.0001 CA1.11764.0001 CA1.11798.0001 CA1.11844.0001 CA1.11845.0001 CA1.11846.0001 CA1.12668.0001 CA1.13124.0001 CA1.13187.0001 CA1.13724.0001 CA1.13725.0001 CA1.13726.0001 CA1.13727.0001 CA1.13728.0001 CA1.13748.0001 CA1.13792.0001 CA1.13793.0001 CA1.13794.0001 CA1.13894.0001 CA1.13895.0001 CA1.15035.0001 CA1.15036.0001 CA1.15037.0001 CA1.15326.0001 CA1.15327.0001 CA1.15341.0001 CA1.15388.0001 CA1.15389.0001 CA1.15423.0001 CA1.15424.0001 CA1.15445.0001 CA1.15552.0001 CA1.15647.0001 CA1.15683.0001 CA1.15684.0001 CA1.15696.0001 CA1.16464.0001 CA1.16485.0001
Precoated Glass-Backed TLC Plates, MilliporeSigma
TLC Plates
For analytical TLC, high-performance analytical TLC, or preparative TLC
F254 is a fluorescent indicator with a 254nm excitation wavelength. F254s is an acid-stable fluorescent indicator with a 254nm excitation wavelength. F254+366 is a fluorescent indicator with 254nm and 366nm excitation wavelengths. RP-18 is reversed phase C18 (with an octadecylsilyl chain). W is for water tolerant, wettable layer.
The sorbents used on these plates are carefully standardized and controlled. Each is characterized by narrow particle size and pore volume distribution, standardized surface area andMore Product Information