Lauryl Sulfate Broth with MUG, Fluorocult, Dehydrated Culture Media, MilliporeSigma
Supplier: MilliporeSigma
1.12588.0500
CA1.12588.0500EA 2764.54 CAD
CA1.12588.0500
Lauryl Sulfate Broth with MUG, Fluorocult, Dehydrated Culture Media, MilliporeSigma
Media for Microbiology Dehydrated Media for Microbiology
Media is granulated.
  • Granulation preserves homogeneity
  • Easy to weigh out
  • Dissolves faster than powdered media
Granulation preserves homogeneity by capturing the media’s original component distribution in each granule and preventing settling out. Dangers associated with inhalation of fine dust and toxic substances are significantly reduced with granulated media. It is easier to weigh out and dissolves faster than powdered media.