HMDS (1,1,1,3,3,3-hexamethyldisilazane) for gas chromatography, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ100

1.12186.0100
CA1.12186.0100CS 45131 CAD
CA1.12186.0100
HMDS (1,1,1,3,3,3-hexamethyldisilazane) for gas chromatography, Supelco®
HMDS (1,1,1,3,3,3-Hexamethyldisilazane)
Formula: (CH₃)₃SiNHSi(CH₃)₃
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: –82 °C
Density: 0.7742 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00008259
CAS Number: 999-97-3
EINECS: 213-668-5
UN: 3286
ADR: 3 (6.1, 8),II
REACH: 01-2119438176-38
Merck Index: 13,04708
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce35a38a4179292c4ff37b2f3093018738545e6e&vid=1693ad54a45ca3ae50a06056893e8438d1002549&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) decomposes
Melting Point -82 °C
Molar Mass 161.39 g/mol
Boiling Point 126 °C (1013 mbar)
Vapor Pressure 20 hPa (20 °C)
Flash Point 6 °C
Explosion Limit 0.8 - 25.9 %(V)
Density 0.774 g/cm3 (20 °C)
Ignition Point 325 °C
Viscosity kinematic 0.9 mm2/s (25 °C)
Hazard Class 3
Hazard Class Description Flammable liquid

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR