Sulfuric acid ≥98.0% (by alkalimetry), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ300

1.12080.1000 1.12080.2500 1.12080.2510 1.12080.9025
CA1.12080.1000CS 583 CAD
CA1.12080.1000 CA1.12080.2500 CA1.12080.2510 CA1.12080.9025
Sulfuric acid ≥98.0% (by alkalimetry), EMSURE® for analysis, Supelco®
Sulfuric acid

For analysis EMSURE®.


Packaging: Safebreak is a robust glass bottle coated with polyethylene (PE). Should the bottle fall and break, all acid and glass splinters are safely retained within the polyethylene coating, thus protecting you from any risk.

Formula: H₂SO₄
Boiling Pt: ~290 °C (lit.)
Melting Pt: –20 °C
Density: 1.840 g/ml (25°C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Merck Index: 14,08974

Order Now

Specification Test Results

Ag (Silver) ≤ 0.000002 %
Al (Aluminium) ≤ 0.000005 %
As (Arsenic) ≤ 0.000001 %
Assay (alkalimetric) ≥ 98.0 %
Ba (Barium) ≤ 0.000005 %
Be (Beryllium) ≤ 0.000001 %
Bi (Bismuth) ≤ 0.00001
Ca (Calcium) ≤ 0.00002 %
Cd (Cadmium) ≤ 0.000002 %
Chloride (Cl) ≤ 0.00001 %
Co (Cobalt) ≤ 0.000001 %
Colour ≤ 10 Hazen
Cr (Chromium) ≤ 0.000005 %
Cu (Copper) ≤ 0.000001 %
Date of expiry see product label
Fe (Iron) ≤ 0.00001
Ge (Germanium) ≤ 0.000005 %
K (Potassium) ≤ 0.00001 %
Li (Lithium) ≤ 0.000001 %
Mg (Magnesium) ≤ 0.000005 %
Mn (Manganese) ≤ 0.000001 %
Mo (Molybdenum) ≤ 0.000005 %
Na (Sodium) ≤ 0.00005 %
NH₄ (Ammonium) ≤ 0.0002 %
Ni (Nickel) ≤ 0.000002 %
Nitrate (NO₃) ≤ 0.00002 %
Pb (Lead) ≤ 0.000002 %
Phosphate (PO₄) ≤ 0.00005 %
Residue on ignition ≤ 0.0005 %
Se (Selenium) ≤ 0.0001 %
Sr (Strontium) ≤ 0.000002 %
Substances reducing potassium permanganate (as SO₂) ≤ 0.0002 %
Ti (Titanium) ≤ 0.00001
Tl (Thallium) ≤ 0.000005 %
V (Vanadium) ≤ 0.000001 %
Zn (Zinc) ≤ 0.000005 %
Zr (Zirconium) ≤ 0.00001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR