Boron, Sigma-Aldrich®

Supplier: MilliporeSigma

1.12070.0010
CA1.12070.0010EA 145 CAD
CA1.12070.0010
Boron, Sigma-Aldrich®
Boron
Formula: B
MW: 10.81 g/mol
MDL Number: MFCD00134034
CAS Number: 7440-42-8
UN: 3178
ADR: 4.1 III
Merck Index: 13,01333

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR