Silver nitrate 0.05 mol/l (0.05 N) in aqueous solution, Titripur® volumetric solution, Supelco®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ100

1.11718.1000
CA1.11718.1000CS 835 CAD
CA1.11718.1000
Silver nitrate 0.05 mol/l (0.05 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Silver nitrate
Formula: AgNO₃
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 3082
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR