Peroxide Test Method: Colorimetric With Test Strips 100 - 200 - 400 - 600 - 800 - 1000 mg/L H₂O₂ MQuant™, MilliporeSigma
Supplier: MilliporeSigma
1.10337.0001
CA1.10337.0001EA 201 CAD
CA1.10337.0001
Peroxide Test Method: Colorimetric With Test Strips 100 - 200 - 400 - 600 - 800 - 1000 mg/L H₂O₂ MQuant™, MilliporeSigma
Environmental Test Kits
The peroxide test is suitable for checking the current concentration of diluted peroxide solutions to ensure the disinfectant effect.
MilliporeSigma # 1.10337.0001 replaces and is identical to MilliporeSigma # 10337-1