Sulfuric acid 0.5 M concentrated aqueous solution, Titrisol®, Supelco®

Supplier: MilliporeSigma
Danger

1.09981.0001
CA1.09981.0001CS 1195 CAD
CA1.09981.0001
Sulfuric acid 0.5 M concentrated aqueous solution, Titrisol®, Supelco®
Sulfuric acid
Formula: H₂SO₄
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,

Order Now

Specification Test Results

Solubility (20 °C) soluble,(caution ! development of heat)
Boiling Point 135 °C
Density 1.29 g/cm3 (20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR