Hydrochloric acid 0.01 M concentrated aqueous solution, Titrisol®, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

1.09974.0001
CA1.09974.0001CS 2197 CAD
CA1.09974.0001
Hydrochloric acid 0.01 M concentrated aqueous solution, Titrisol®, Supelco®
Hydrochloric acid
Formula: HCl
MW: 36.46
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.00 g/cm3 (20 °C)
pH 1 (H2O) (undiluted)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR