p-Xylene reference substance for gas chromatography, Supelco®

Supplier: MilliporeSigma
Danger

1.09799.0005
CA1.09799.0005CS 101454 CAD
CA1.09799.0005
p-Xylene reference substance for gas chromatography, Supelco®
p-Xylene
Formula: C₆H₄(CH₃)₂
Boiling Pt: 138.3 °C (1013 hPa)
Melting Pt: 13 °C
Density: 1.096 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00008556
CAS Number: 106-42-3
EINECS: 203-396-5
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8b04563a5941cefd103ebd3048bcf8486f2bdac&vid=e5db83ad1f0cb6a583610c58cea291dcbe1be1f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.2 g/l (25 °C)
Melting Point 13.3 °C
Molar Mass 106.17 g/mol
Boiling Point 138.4 °C (1013 hPa)
Vapor Pressure 8.7 hPa (20 °C)
Flash Point 27.2 °C
Explosion Limit 1.1 - 7 %(V)
Density 0.86 g/cm3 (20 °C)
Ignition Point 540 °C
Saturation concentration 38 g/m3 (20 °C) Air
Evaporation number 13.5
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR