Certipur® TOC standard solution acc. to EN 1484-H3/DIN 38409-H3 as potassium hydrogen phthalate in water, stabilized
Supplier: MilliporeSigma
1.09017.0100
CA1.09017.0100CS 3023 CAD
CA1.09017.0100
Certipur® TOC standard solution acc. to EN 1484-H3/DIN 38409-H3 as potassium hydrogen phthalate in water, stabilized
Standards Environmental Analysis Standards
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST (SRM 84L)
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors
Caution: Store at +15 to +25 °C.