Hydrogen peroxide 35%, Technical Grade, Supelco®

Supplier: MilliporeSigma
Danger

1.08556.9025
CA1.08556.9025EA 1179 CAD
CA1.08556.9025
Hydrogen peroxide 35%, Technical Grade, Supelco®
Hydrogen peroxide
Formula: H₂O₂
Boiling Pt: ∼107 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 (8),II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -24 °C
Boiling Point 110 °C (1013 hPa)
Vapor Pressure 20 hPa (20 °C)
Density 1.13 g/cm3 (20 °C)
pH 2 - 4 (H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR