3,3',5,5'-Tetramethylbenzidine (TMB) solution for the detection of chlorine, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine

1.08475.1000
CA1.08475.1000CS 1087 CAD
CA1.08475.1000
3,3',5,5'-Tetramethylbenzidine (TMB) solution for the detection of chlorine, Supelco®
3,3',5,5'-Tetramethylbenzidine (TMB)
Formula: C₁₆H₂₀N₂
MW: 240.35 g/mol
Boiling Pt: 402
MDL Number: MFCD00007748
CAS Number: 54827-17-7
UN: 3264
ADR: 8 II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.023 g/cm3
Hazard Class 8
Hazard Class Description Corrosive material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR