2,3,5-Triphenyltetrazolium chloride (TTC) ≥98% (by perchloric acid titration) for microbiology

Supplier: MilliporeSigma
Danger

1.08380.0100
CA1.08380.0100EA 1331.99 CAD
CA1.08380.0100
2,3,5-Triphenyltetrazolium chloride (TTC) ≥98% (by perchloric acid titration) for microbiology
2,3,5-Triphenyltetrazolium chloride
Formula: C₁₉H₁₅ClN₄
Melting Pt: 243 °C (dec.)
MDL Number: MFCD00011963
CAS Number: 298-96-4
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Assay (perchloric acid titration, calculated on dried substance) ≥98%
Identity (UV/VIS-Spectrum) Passes Test
Absorption maximum λmax. (water) 245 - 248 nm
Spec. Absorptivity A 1%/1cm (λmax; 0,01 g/L; water; calculated on dried substance) ≥770
TLC-Test Passes Test
Loss on drying (70 °C; ≤1 hPa) ≤0.5%
Suitability for testing the germinability of seeds Passes Test
Suitability for microbiology Passes Test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR