4-Aminoantipyrine, GR for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone, 4-Aminophenazone

1.07293.0010 1.07293.0100
CA1.07293.0010CS 10583 CAD
CA1.07293.0010 CA1.07293.0100
4-Aminoantipyrine, GR for analysis, Supelco®
4-Aminoantipyrine
Formula: C₁₁H₁₃N₃O
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00003145
CAS Number: 83-07-8
EINECS: 201-452-3
Merck Index: 13,00596

Order Now

Specification Test Results

Solubility 56 g/l (20 °C)
Melting Point 107 - 109 °C
Molar Mass 203.24 g/mol
Bulk Density 430 kg/m3
pH 7.1 (100 g/l, H2O, 20 °C) (slurry)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR