Sodium dihydrogen phosphate dihydrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: Phosphoric acid (sodium hydrate), Sodium phosphate monobasic dihydrate, Sodium dihydrogen orthophosphate dihydrate

1.06342.0250 1.06342.1000 1.06342.2500
CA1.06342.0250EA 70 CAD
CA1.06342.0250 CA1.06342.1000 CA1.06342.2500
Sodium dihydrogen phosphate dihydrate, EMSURE® for analysis, Supelco®
Sodium dihydrogen phosphate dihydrate
Formula: NaH₂PO₄·2H₂O
Boiling Pt: ∼650 °C (1013 hPa)
Melting Pt: 60 °C
Density: 1.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149209
CAS Number: 13472-35-0
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Order Now

Specification Test Results

Solubility 850 g/l (20 °C)
Melting Point 60 °C
Molar Mass 156.02 g/mol
Bulk Density 1000 kg/m3
Density 1.915 g/cm3 (20 °C)
pH 4.5 (12 g/l, H2O, 25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR