Magnesium chloride hexahydrate 99.0-101.0% (by complexometric analysis), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ300

1.05833.0250 1.05833.1000 1.05833.5000 1.05833.9025
CA1.05833.0250EA 188 CAD
CA1.05833.0250 CA1.05833.1000 CA1.05833.5000 CA1.05833.9025
Magnesium chloride hexahydrate 99.0-101.0% (by complexometric analysis), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Magnesium chloride hexahydrate

EMSURE® (formerly GR) is a premium grade of reagents for wet chemical analysis, adhering to ACS, ISO, and/or Reag. Ph.Eur quality specifications.


n North America 105833.0250 replaces but has a far superior specification to MX0045-2
In North America 105833.0250 replaces MX0045-1 but has a far superior specification and a different pack size
In North America 105833.1000 replaces but has a far superior specification and different pack size to MX0045-1
In North America 105833.1000 replaces but has a far superior specification to MX0045-4
In North America 105833.9025 replaces MX0045-20 but has a far superior specification and a different pack size


Certifications: Corresponds to ACS, ISO, Reag. Ph Eur

Formula: MgCl₂·6H₂O
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C (dec.)
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Specification Test Results

Assay (complexometric) 99.0 - 101.0 %
Identity conforms
Appearance of solution passes test
Insoluble matter ≤ 0.005 %
pH-value (5 %; water) 5.0 - 6.5
Acidity or alkalinity passes test
Bromide (Br) ≤ 0.05 %
Nitrate (NO₃) ≤ 0.001 %
Phosphate (PO₄) ≤ 0.0005 %
Sulfate (SO₄) ≤ 0.002 %
Total nitrogen (N) ≤ 0.0002 %
Heavy metals (as Pb) ≤ 0.0005 %
Al (Aluminium) ≤ 0.0002 %
As (Arsenic) ≤ 0.0002 %
Ba (Barium) ≤ 0.002 %
Ca (Calcium) ≤ 0.003 %
Cu (Copper) ≤ 0.0005 %
Fe (Iron) ≤ 0.0005 %
K (Potassium) ≤ 0.001 %
Mn (Manganese) ≤ 0.0005 %
Na (Sodium) ≤ 0.001 %
NH₄ (Ammonium) ≤ 0.002 %
Pb (Lead) ≤ 0.0005 %
Sr (Strontium) ≤ 0.005 %
Water 51.0 - 55.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR