SABOURAUD 4% Dextrose Agar, MilliporeSigma
Supplier: MilliporeSigma
1.05438.5000
CA1.05438.5000EA 1360 CAD
CA1.05438.5000
SABOURAUD 4% Dextrose Agar, MilliporeSigma
Media for Microbiology Dehydrated Media for Microbiology
In North America 1.05438.5000 replaces and is identical to 1.05438.5007
  • Clear, yellowish-brown appearance
  • Fair to very good growth in Trichophyton mentagrophytes ATCC 18748
  • Fair to good growth in Trichophyton rubrum ATCC 28188
  • Fair to good growth in Trichophyton ajelloi ATCC 28454
  • Fair to very good growth in Microsporum gallinae ATCC 12108
  • Good to very good growth in Microsporum canis ATCC 36299
  • Good to very good growth in Geotrichum candidum DSM 1240
  • Good to very good growth in Penicillium commune ATCC 10428