Potassium thiocyanate ≥99.0% (silver nitrate titration), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.05125.0250 1.05125.1000 1.05125.9050
CA1.05125.0250EA 276 CAD
CA1.05125.0250 CA1.05125.1000 CA1.05125.9050
Potassium thiocyanate ≥99.0% (silver nitrate titration), EMSURE® for analysis, Supelco®
Potassium thiocyanate

1.05125.0250 is offered as an alternative to PX1630-3. Different Pack Size, Same Specifications.
1.05125.1000 is offered as an alternative to PX1630-1. Different Pack Size, Same Specifications.
1.05125.1000 is offered as an alternative to PX1630-5. Different Pack Size, Same Specifications.
1.05125.9050 is offered as an alternative to PX1630-7. different size, different packaging, same chem spec.
1.05125.9050 replaces and is the same as PX1630-20. Part # Change only, Same Pack Size, Same Specifications.

Formula: KSCN
Boiling Pt: >400 °C (1013 hPa) (dec.)
Melting Pt: 177 °C
Density: 1.91 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1
Merck Index: 12,07866

Order Now

Specification Test Results

Solubility 208 g/l (20 °C)
Melting Point 175 °C
Molar Mass 97.18 g/mol
Bulk Density 750 - 1000 kg/m3
Vapor Pressure  - 1 hPa (20 °C)
Density 1.89 g/cm3 (20 °C)
pH 5.3 - 8.5 (50 g/l, H2O, 20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR