Potassium hexacyanoferrate(II) trihydrate 99.0-102.0% (by cerimetric analysis), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

1.04984.0100 1.04984.0500 1.04984.9050
CA1.04984.0100EA 213 CAD
CA1.04984.0100 CA1.04984.0500 CA1.04984.9050
Potassium hexacyanoferrate(II) trihydrate 99.0-102.0% (by cerimetric analysis), EMSURE® for analysis, Supelco®
Potassium hexacyanoferrate (II) trihydrate

1.04984.0500 replaces and is the same as PX1460-1. Part # Change only, Same Pack Size, Same Specifications.
1.04984.0500 is offered as an alternative to PX1460-5. Different Pack Size, Same Specifications.
1.04984.9050 is offered as an alternative to PX1460-7. different size, different packaging, same chem spec.
1.04984.9050 replaces and is the same as PX1460-20. Part # Change only, Same Pack Size, Same Specifications.

Formula: K₄Fe(CN)₆·3H₂O
Melting Pt: >50 °C (–H₂O of crystall.)
Density: 1.889 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167023
CAS Number: 14459-95-1
EINECS: 237-722-2
Merck Index: 12,07793

Order Now

Specification Test Results

Solubility 289 g/l (20 °C)
Melting Point 70 °C Elimination of water of crystallisation
Molar Mass 422.39 g/mol
Bulk Density 950 - 1050 kg/m3
Density 1.85 g/cm3 (20 °C) (anhydrous substance)
pH 9.5 (100 g/l, H2O, 20 °C) (anhydrous substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR