myo-Inositol ≥99% (by HPLC) for microbiology, Millipore®

Supplier: MilliporeSigma

Synonyms: i-Inositol, D-Myo-Inositol, rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaol, Inositol

1.04728.0100
CA1.04728.0100EA 164.19 CAD
CA1.04728.0100
myo-Inositol ≥99% (by HPLC) for microbiology, Millipore®
myo-Inositol
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 291
Melting Pt: 225…227 °C
Storage Temperature: Ambient
MDL Number: MFCD00077932
CAS Number: 87-89-8
Merck Index: 13,05001

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR