Mercury(II) chloride, fine crystals, EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

1.04417.0100
CA1.04417.0100EA 306 CAD
CA1.04417.0100 CA1.04417.1000
Mercury(II) chloride, fine crystals, EMPLURA®, Supelco®
Mercury (II) chloride
Formula: HgCl₂
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 273 °C
Density: 5.44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011041
CAS Number: 7487-94-7
EINECS: 231-299-8
UN: 1624
ADR: 6.1,II
Merck Index: 13,05901

Order Now

Specification Test Results

Assay (complexometric; calculated on dried substance) 99.5 - 100.5 %
Identity passes test
Appearance of solution passes test
Acidity or alkalinity passes test
Mercury(I)-chloride passes test
Loss on drying ≤ 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR