Listeria Selective Supplement (UVM-II) Dehydrated Culture Media, EMD Millipore
Supplier: MilliporeSigma
1.04039.0001
CA1.04039.0001EA 314.19 CAD
CA1.04039.0001
Listeria Selective Supplement (UVM-II) Dehydrated Culture Media, EMD Millipore
Media for Microbiology Dehydrated Media for Microbiology
UVM II supplement for microbiology (1 vial with 13 mg of acriflavine hydrochloride)
  • Granulation preserves homogeneity
  • Easy to weigh out
  • Dissolves faster than powdered media
Granulation preserves homogeneity by capturing the media’s original component distribution in each granule and preventing settling out. Dangers associated with inhalation of fine dust and toxic substances are significantly reduced with granulated media. It is easier to weigh out and dissolves faster than powdered media.
CAS 69235-50-3, chemical formula C₁₄H₁₅Cl₂N₃ + C₁₃H₁₃Cl₂N₃.