Ammonium cerium(IV) nitrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Ammonium ceric nitrate, Ammonium hexanitratocerate (IV), Ceric Ammonium Nitrate, Cerium (IV) ammonium nitrate, Diammonium hexanitratocerate

1.02276.0100 1.02276.1000
CA1.02276.0100CS 12374 CAD
CA1.02276.0100 CA1.02276.1000
Ammonium cerium(IV) nitrate, EMSURE® for analysis, Supelco®
Ammonium cerium (IV) nitrate
Formula: (NH₄)₂Ce(NO₃)₆
MW: 548.22 g/mol
Melting Pt: 107…108 °C
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151121
CAS Number: 16774-21-3
EINECS: 240-827-6
UN: 3085
ADR: 5.1 (8) II
Merck Index: 13,00509

Order Now

Specification Test Results

Solubility 1410 g/l (20 °C)
Melting Point 108 °C
Molar Mass 548.22 g/mol
Bulk Density 1200 kg/m3
Density 2.49 g/cm3
pH 1 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR