Silver sulfate 10 g/L in sulphuric acid (d = 1.84 g/mL) for COD determination, Supelco®

Supplier: MilliporeSigma
Danger

1.01517.1000 1.01517.2500
CA1.01517.1000CS 3975 CAD
CA1.01517.1000 CA1.01517.2500
Silver sulfate 10 g/L in sulphuric acid (d = 1.84 g/mL) for COD determination, Supelco®
Silver sulphate
Formula: Ag₂SO₄
MDL Number: MFCD00003407
CAS Number: 10294-26-5
EINECS: 233-653-7
UN: 3264
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble,(caution ! development of heat)
Density 1.84 g/cm3 (20 °C)
pH (H2O, 20 °C) strongly acid
Hazard Class 8
Hazard Class Description Corrosive material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR