Diethyl ether ≥99.7% (by GC), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.00921.1000 1.00921.2500 1.00921.6010 1.00921.6025 1.00921.6190
CA1.00921.0190EA 11324.43 CAD
CA1.00921.0190 CA1.00921.1000 CA1.00921.2500 CA1.00921.5000 CA1.00921.6010 CA1.00921.6025 CA1.00921.6190 CA1.00921.9190
Diethyl ether ≥99.7% (by GC), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Diethyl ether
Formula: (CH₃CH₂)₂O
Boiling Pt: 34.6 °C (lit.)
Melting Pt: –116 °C (lit.)
Density: 0.706 g/ml (25°C) (lit.)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 69 g/l (20 °C)
Melting Point -116.3 °C
Molar Mass 74.12 g/mol
Boiling Point 34.6 °C (1013 hPa)
Vapor Pressure 587 hPa (20 °C)
Flash Point -40 °C
Explosion Limit 1.7 - 36 %(V)
Density 0.71 g/cm3
Ignition Point 180 °C
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR